Learn More
Cayman Chemical Moclobemide-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 2929883-33-8, 1 mg
SDP
Catalog No. 502950377
Encompass_Preferred
An internal standard for the quantification of moclobemide by GC- or LC-MS
Catalog No. 50-295-0377 Supplier Cayman Chemical Supplier No. 342471 MG
An internal standard for the quantification of moclobemide by GC- or LC-MS