Learn More
Cayman Chemical Moclobemide-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 2929883-33-8, 5 mg
SDP
Catalog No. 502946823
Encompass_Preferred
An internal standard for the quantification of moclobemide by GC- or LC-MS
Catalog No. 50-294-6823 Supplier Cayman Chemical Supplier No. 342475 MG
An internal standard for the quantification of moclobemide by GC- or LC-MS