Learn More
Medchemexpress LLC Moclobemide-d4 | 71320-77-9 | 99.0% | 1 MG
SDP
Catalog No. 5000407962
Encompass_Preferred
Moclobemide-d4 is deuterium labeled Moclobemide. Moclobemide (Ro111163) is a brain-penetrant and reversible monoamine oxidase (MAO-A) inhibitor with an IC50 of 6.061 μM for hMAO-A. It up-regulates proliferation of hippocampal progenitor cells in chronically stressed mice. This product is for research use only and is not sold to patients.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-07962 Supplier Medchemexpress LLC Supplier No. HYB0534S11MG
Moclobemide-d4 is deuterium labeled Moclobemide. Moclobemide (Ro111163) is a brain-penetrant and reversible monoamine oxidase (MAO-A) inhibitor with an IC50 of 6.061 μM for hMAO-A. It up-regulates proliferation of hippocampal progenitor cells in chronically stressed mice. This product is for research use only and is not sold to patients.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS