Learn More
Medchemexpress LLC Moclobemide-d4 | 71320-77-9 | 99.0% | 100 UG
SDP
Catalog No. 5000407570
Encompass_Preferred
Moclobemide-d4 is a deuterium-labeled form of Moclobemide, a brain-penetrant and reversible monoamine oxidase (MAO-A) inhibitor. It has shown to up-regulate proliferation of hippocampal progenitor cells in chronically stressed mice and is used as a tracer or internal standard in quantitative analysis. Stable heavy isotopes, like deuterium, are incorporated into drug molecules for quantitation during drug development and can influence pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Affects pharmacokinetic and metabolic profiles of drugs
  • Up-regulates proliferation of hippocampal progenitor cells in chronically stressed mice
Catalog No. 50-004-07570 Supplier Medchemexpress LLC Supplier No. HYB0534S1100UG
Moclobemide-d4 is a deuterium-labeled form of Moclobemide, a brain-penetrant and reversible monoamine oxidase (MAO-A) inhibitor. It has shown to up-regulate proliferation of hippocampal progenitor cells in chronically stressed mice and is used as a tracer or internal standard in quantitative analysis. Stable heavy isotopes, like deuterium, are incorporated into drug molecules for quantitation during drug development and can influence pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Affects pharmacokinetic and metabolic profiles of drugs
  • Up-regulates proliferation of hippocampal progenitor cells in chronically stressed mice