Learn More
Medchemexpress LLC Moclobemide-d4 | >99.0% | 5 MG
SDP
Catalog No. 5000405571
Encompass_Preferred
Moclobemide-d4 is a deuterium-labeled version of Moclobemide. Moclobemide (Ro111163) is identified as a brain-penetrant and reversible inhibitor of monoamine oxidase (MAO-A), with an IC50 of 6.061 μM for hMAO-A. It has been observed to up-regulate the proliferation of hippocampal progenitor cells in chronically stressed mice.
  • Deuterium labeled compound
  • Primarily intended for research purposes
  • Can be utilized as a tracer or an internal standard for quantitative analysis through methods like NMR, GC-MS, or LC-MS
  • Useful in pharmacokinetic studies
Catalog No. 50-004-05571 Supplier Medchemexpress LLC Supplier No. HYB0534S15MG
Moclobemide-d4 is a deuterium-labeled version of Moclobemide. Moclobemide (Ro111163) is identified as a brain-penetrant and reversible inhibitor of monoamine oxidase (MAO-A), with an IC50 of 6.061 μM for hMAO-A. It has been observed to up-regulate the proliferation of hippocampal progenitor cells in chronically stressed mice.
  • Deuterium labeled compound
  • Primarily intended for research purposes
  • Can be utilized as a tracer or an internal standard for quantitative analysis through methods like NMR, GC-MS, or LC-MS
  • Useful in pharmacokinetic studies