Learn More
Selleck Chemical LLC Moclobemide (Ro 111163) 10mg 71320-77-9
SDP
Supplier:  Selleck Chemical LLC S321210MG
Encompass_Preferred
Moclobemide is a MAO-A (5-HT) inhibitor with IC50 of 6.1 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3507