Learn More
Apexbio Technology LLC Moclobemide (Ro 111163) 71320-77-9 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062258
Encompass_Preferred
Moclobemide (Ro 111163 CAS 71320-77-9) is a reversible inhibitor of monoamine oxidase A (MAO-A) with a reported IC50 of approximately 10 M In vitro studies using rat brain homogenate indicate that it selectively targets MAO-A over MAO-B though with moderate potency Animal experiments demonstrate up to 80% inhibition of MAO-A activity in brain and liver tissues upon administration Prolonged high-dose exposure to Moclobemide alters adrenergic receptor profiles by downregulating -adrenergic receptors and enhancing affinity of 1-adrenergic agonist binding Due to its MAO-A specificity this compound serves as a prototype reversible inhibitor (RIMA) with relevance in antidepressant drug research
Catalog No. 50-000-62258 Supplier Apexbio Technology LLC Supplier No. A43705.1
 Restricted Item
Add to Cart
Add to Cart
Moclobemide (Ro 111163 CAS 71320-77-9) is a reversible inhibitor of monoamine oxidase A (MAO-A) with a reported IC50 of approximately 10 M In vitro studies using rat brain homogenate indicate that it selectively targets MAO-A over MAO-B though with moderate potency Animal experiments demonstrate up to 80% inhibition of MAO-A activity in brain and liver tissues upon administration Prolonged high-dose exposure to Moclobemide alters adrenergic receptor profiles by downregulating -adrenergic receptors and enhancing affinity of 1-adrenergic agonist binding Due to its MAO-A specificity this compound serves as a prototype reversible inhibitor (RIMA) with relevance in antidepressant drug research