Learn More
Medchemexpress LLC Mocravimod hydrochloride | 509088-69-1 | 99.99% | 480.45 | 1 ML
SDP
Catalog No. 5000351940
Encompass_Preferred
Mocravimod hydrochloride is an orally active sphingosine-1-phosphate receptor (S1PR) modulator. It works by blocking the signal required by T cells to egress from lymph nodes and other lymphoid organs, preferentially binding to S1PR1 over S1PR2 and S1PR3 in cardiomyocytes. This compound retains T cell effector function and is suitable for various research applications.
  • Significantly lowers the concentration of reactive oxygen species (ROS).
  • Prevents mitochondrial permeability transition pore opening.
  • Boosts mitochondrial membrane potential (MMP).
  • Increases phosphorylation of AKT, EKR, GSK-3β, JAK2, and STAT3.
  • Retains T cell effector function.
  • Used for the study of acute myelogenous leukemia, diabetes, and Myocardial Ischemia-Reperfusion Injury (MIRI).
Catalog No. 50-003-51940 Supplier Medchemexpress LLC Supplier No. HY1366010MM/1ML
Mocravimod hydrochloride is an orally active sphingosine-1-phosphate receptor (S1PR) modulator. It works by blocking the signal required by T cells to egress from lymph nodes and other lymphoid organs, preferentially binding to S1PR1 over S1PR2 and S1PR3 in cardiomyocytes. This compound retains T cell effector function and is suitable for various research applications.
  • Significantly lowers the concentration of reactive oxygen species (ROS).
  • Prevents mitochondrial permeability transition pore opening.
  • Boosts mitochondrial membrane potential (MMP).
  • Increases phosphorylation of AKT, EKR, GSK-3β, JAK2, and STAT3.
  • Retains T cell effector function.
  • Used for the study of acute myelogenous leukemia, diabetes, and Myocardial Ischemia-Reperfusion Injury (MIRI).