Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions Bromine Index Number Titration Solvent (ASTM D5776-14a) (4L)
SDP

Catalog No. NC9553701
Encompass_Preferred

Bromine Index Number Titration Solvent (ASTM D5776-14a) (4L)

Catalog No. NC9553701 Supplier Aqua Solutions Supplier No. 13654L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bromine Index Number Titration Solvent (ASTM D5776-14a) (4L)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.