Learn More
NEOGEN Modified Giolitti & Cantoni Broth, 5 kg
SDP
Catalog No. 502015676
Encompass_Preferred
Modified Giolitti and Cantoni Broth (ISO) is used for the detection and enumeration of coagulase- positive staphylococci from food and animal feeding stuffs using the Most Probable Number (MPN) technique according to ISO 6888-3:2003. This medium is not intended for use in the diagnosis of disease or other conditions in humans.Originally described by Giolitti and Cantoni as a medium for the enrichment of staphylococci from foodstuffs, Mossel later applied the medium to use with samples from dried milk and infant food. Optimized for use in samples where staphylococci may be stressed and/or in low numbers, growth of the target organisms is promoted by sodium pyruvate, Glycine and the high concentration of mannitol.For more information visit https://www.neogen.com/solutions/Microbiology/modified-giolitti-and-cantoni-broth
Catalog No. 50-201-5676 Supplier NEOGEN Supplier No. 700004581
Modified Giolitti and Cantoni Broth (ISO) is used for the detection and enumeration of coagulase- positive staphylococci from food and animal feeding stuffs using the Most Probable Number (MPN) technique according to ISO 6888-3:2003. This medium is not intended for use in the diagnosis of disease or other conditions in humans.Originally described by Giolitti and Cantoni as a medium for the enrichment of staphylococci from foodstuffs, Mossel later applied the medium to use with samples from dried milk and infant food. Optimized for use in samples where staphylococci may be stressed and/or in low numbers, growth of the target organisms is promoted by sodium pyruvate, Glycine and the high concentration of mannitol.For more information visit https://www.neogen.com/solutions/Microbiology/modified-giolitti-and-cantoni-broth