Learn More
Sigma Aldrich Fine Chemicals Biosciences Hematoxylin Solution, Harris Modified, 1412-19-7, MFCD00078111, 500 mL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences HHS16500ML
Encompass
Molecular Formula: C16 H14 O6, Molecular Weight: 302.281. General purpose nuclear stain, regressive type. Used with hematoxylin and eosin staining.
Catalog No. NC0310239