Learn More
Medchemexpress LLC Modaline sulfate | 2856-75-9 | 99.4% | 50 MG
SDP
Catalog No. 5000413885
Encompass_Preferred
Modaline sulfate is a monoamine oxidase (MAO) inhibitor that has been investigated for its potential in treating depression. It is provided as a solid with a light yellow to green-yellow appearance.
  • MAO inhibitor
  • Used in the treatment of depression
  • Available in solid form
  • Soluble in DMSO (≥ 31 mg/mL)
  • For research use only
Catalog No. 50-004-13885 Supplier Medchemexpress LLC Supplier No. HYB108350MG
Modaline sulfate is a monoamine oxidase (MAO) inhibitor that has been investigated for its potential in treating depression. It is provided as a solid with a light yellow to green-yellow appearance.
  • MAO inhibitor
  • Used in the treatment of depression
  • Available in solid form
  • Soluble in DMSO (≥ 31 mg/mL)
  • For research use only