Learn More
Grainger LISTA Modular Drawer Cabinet, Steel, 7 Drawers, 114 Compartments, Gray, 1 ea
Catalog No. 502769539
Encompass_Preferred
This modular drawer cabinet is designed for high-density storage and can be integrated with other modular cabinets and base shelf units to create a custom storage system. The stationary, welded steel cabinet features 7 drawers providing 114 compartments for organized tool or component storage, finished with a durable powder coat.
  • Features 7 drawers with a 440 lb. load capacity per drawer.
  • Includes 114 total compartments with adjustable dividers and partitions.
  • Constructed from heavy-duty steel with a powder coated finish.
  • Equipped with full width recessed handles.
  • Includes keys for secure locking.
  • Stackable design for increased storage density.
  • Stationary base frame configuration.
Catalog No. 50-276-9539 Supplier Grainger Supplier No. 13P572
This modular drawer cabinet is designed for high-density storage and can be integrated with other modular cabinets and base shelf units to create a custom storage system. The stationary, welded steel cabinet features 7 drawers providing 114 compartments for organized tool or component storage, finished with a durable powder coat.
  • Features 7 drawers with a 440 lb. load capacity per drawer.
  • Includes 114 total compartments with adjustable dividers and partitions.
  • Constructed from heavy-duty steel with a powder coated finish.
  • Equipped with full width recessed handles.
  • Includes keys for secure locking.
  • Stackable design for increased storage density.
  • Stationary base frame configuration.