Learn More
Arrow Engineering Direct Drive Overhead Stirrer, Electric Model 1200, 120V 60HZ
Catalog No. NC1603805
Encompass_Preferred
Known for its durability and superior performance, the Arrow Mixing Products Model 1200 is an essential asset for professionals who require reliable and efficient mixing solutions. This heavy-duty 1/9HP mixer comes equipped with an adjustable speed dial, allowing operators to control the mixing speed seamlessly for precise processing with a speed range of 50-1200 RPM's. The Model 1200 comes standard with a 3/8" dia x 12" propeller shaft, 2-1/2"dia 4 Blade propeller & aluminum coupling.
Catalog No. NC1603805 Supplier Arrow Engineering Supplier No. ARROW 1200
Known for its durability and superior performance, the Arrow Mixing Products Model 1200 is an essential asset for professionals who require reliable and efficient mixing solutions. This heavy-duty 1/9HP mixer comes equipped with an adjustable speed dial, allowing operators to control the mixing speed seamlessly for precise processing with a speed range of 50-1200 RPM's. The Model 1200 comes standard with a 3/8" dia x 12" propeller shaft, 2-1/2"dia 4 Blade propeller & aluminum coupling.