Learn More
J Kem Scientific Inc MODEL 150 CONTROLLER K SENSOR
SDP
Catalog No. NC3343380
Encompass
Model 150 Controller only with K-type temperature sensor. An ideal controller for applications that don’t require the precise regulation of J-KEM’s 200-Series controllers. This compact unit packs 1200 watts of power, sufficient for 5 L heating mantles, many ovens, and other devices. Built with J-KEM’s state-of-the-art PID process controller, but does not contain J-KEM’s power control computer.
Catalog No. NC3343380 Supplier J Kem Scientific Inc Supplier No. 150K
Model 150 Controller only with K-type temperature sensor. An ideal controller for applications that don’t require the precise regulation of J-KEM’s 200-Series controllers. This compact unit packs 1200 watts of power, sufficient for 5 L heating mantles, many ovens, and other devices. Built with J-KEM’s state-of-the-art PID process controller, but does not contain J-KEM’s power control computer.