Learn More
Sentry Air Systems Model 200 99.97% HEPA 2 Pack
SDP
Supplier:  Sentry Air Systems SS200HF
Encompass
HEPA Filters Efficiency: up to 99.97% efficiency on particles down to 0.3 microns in size. MERV Rating: MERV 17 set by the US Department of Energy to ensure high-quality filtration standards. HEPA filters provide high-quality capture and containment of particulates emitted from applications. The Model 200 HEPA filters (2 pack) fit Sentry Air Systems Model 200 Fume Extractors, Ductless Fume Hoods, and Portable Clean Rooms. 100% fire retardant and handcrafted with specialty purpose fiberglass material. Backed by multi-level quality assurance.
Catalog No. NC1320468