Learn More
Sentry Air Systems Model 300 Activated Carbon/Formaldehyde Filter - 10 lb.
SDP
Supplier:  Sentry Air Systems SS310FORM
Encompass
Model 300 Formaldehyde Filter features 10 lbs of heavy-duty activated carbon treated for aldehyde removal. Fits Model 300 systems.
Catalog No. NC2034264