Learn More
Electron Microscopy Sciences U-tek Model 312 Mini Mailers for Temperature SensitiveShipments
SDP
Catalog No. 5033830
Encompass_Preferred
Compact protection for temperature sensitive shipments, ideal for serums, tissues, and reagents. Accommodate tubes, vials, ampoules, small bottles, and tissue flasks with space for refrigerants. Insulating, thick wall polystyrene foam construction.
Catalog No. 50-338-30 Supplier Electron Microscopy Sciences Supplier No. 6152012
Compact protection for temperature sensitive shipments, ideal for serums, tissues, and reagents. Accommodate tubes, vials, ampoules, small bottles, and tissue flasks with space for refrigerants. Insulating, thick wall polystyrene foam construction.