Learn More
Electron Microscopy Sciences U-tek Model 335 Mini Mailers
SDP
Catalog No. 5033824
Encompass_Preferred
Compact protection for temperature sensitive shipments, ideal for serums, tissues, and reagents. Accommodate tubes, vials, ampoules, small bottles, and tissue flasks with space for refrigerants. Insulating, thick wall polystyrene foam construction. Holds specimen tubes, vials, swabs.
Catalog No. 50-338-24 Supplier Electron Microscopy Sciences Supplier No. 6152235
Compact protection for temperature sensitive shipments, ideal for serums, tissues, and reagents. Accommodate tubes, vials, ampoules, small bottles, and tissue flasks with space for refrigerants. Insulating, thick wall polystyrene foam construction. Holds specimen tubes, vials, swabs.