Learn More
Electron Microscopy Sciences ThermoSafe Model 340 Mailing Sleeves
SDP
Catalog No. 5033812
Encompass_Preferred
Sturdy 18-point white paperboard sleeves increase the cost-effectiveness of foam mailers. Reverse tuck closures enable foam mailers to withstand multiple trips. Fits Foam Mailer 364, 339, and 394.
Catalog No. 50-338-12 Supplier Electron Microscopy Sciences Supplier No. 6151640
Sturdy 18-point white paperboard sleeves increase the cost-effectiveness of foam mailers. Reverse tuck closures enable foam mailers to withstand multiple trips. Fits Foam Mailer 364, 339, and 394.