Learn More
Electron Microscopy Sciences ThermoSafe Model 356 LabMailer
SDP
Catalog No. 5033801
Encompass_Preferred
Maximum breakage protection for known non-hazadous blood, serum, tissue, and other specimens. Accommodates  VacuTainer  Microtainer and other collection / transport tubes. Lightweight polystyrene foam construction saves postage. Reusable and recyclable. Holds one tube, 110 x 16ml, 357 Mailing Sleeve.
Catalog No. 50-338-01 Supplier Electron Microscopy Sciences Supplier No. 6151556
Maximum breakage protection for known non-hazadous blood, serum, tissue, and other specimens. Accommodates  VacuTainer  Microtainer and other collection / transport tubes. Lightweight polystyrene foam construction saves postage. Reusable and recyclable. Holds one tube, 110 x 16ml, 357 Mailing Sleeve.