Learn More
Electron Microscopy Sciences ThermoSafe Model 369 LabMailer
SDP
Catalog No. 5033798
Encompass_Preferred
Maximum breakage protection for known non-hazadous blood, serum, tissue, and other specimens. Accommodates  VacuTainer  Microtainer and other collection / transport tubes. Lightweight polystyrene foam construction saves postage. Reusable and recyclable. Holds 8 slides, 370 Mailing Sleeve.
Catalog No. 50-337-98 Supplier Electron Microscopy Sciences Supplier No. 6151569
Maximum breakage protection for known non-hazadous blood, serum, tissue, and other specimens. Accommodates  VacuTainer  Microtainer and other collection / transport tubes. Lightweight polystyrene foam construction saves postage. Reusable and recyclable. Holds 8 slides, 370 Mailing Sleeve.