Learn More
Electron Microscopy Sciences U-tek Model 395 Mini Mailers
SDP
Catalog No. 5033823
Encompass_Preferred
Compact protection for temperature sensitive shipments, ideal for serums, tissues, and reagents. Accommodate tubes, vials, ampoules, small bottles, and tissue flasks with space for refrigerants. Insulating, thick wall polystyrene foam construction. Holds 2 oz Boston round bottles.
Catalog No. 50-338-23 Supplier Electron Microscopy Sciences Supplier No. 6152295
Compact protection for temperature sensitive shipments, ideal for serums, tissues, and reagents. Accommodate tubes, vials, ampoules, small bottles, and tissue flasks with space for refrigerants. Insulating, thick wall polystyrene foam construction. Holds 2 oz Boston round bottles.