Learn More
Sentry Air Systems Model 400 Magnehelic Gauge
SDP
Catalog No. NC2161059
Encompass
The Magnehelic Gauge monitors the filter saturation for Model 400 units. This item displays the static pressure on the filter in inches of water column.
This item is not returnable. View return policy
Catalog No. NC2161059 Supplier Sentry Air Systems Supplier No. SS400MAG
The Magnehelic Gauge monitors the filter saturation for Model 400 units. This item displays the static pressure on the filter in inches of water column.