Learn More
Electron Microscopy Sciences ThermoSafe Model 407 LabMailer
SDP
Catalog No. 5033805
Encompass_Preferred
Maximum breakage protection for known non-hazadous blood, serum, tissue, and other specimens. Accommodates  VacuTainer  Microtainer and other collection / transport tubes. Lightweight polystyrene foam construction saves postage. Reusable and recyclable. Holds 4 tubes, 110 x 19ml, 8 slides, 408 Mailing Sleeve.
Catalog No. 50-338-05 Supplier Electron Microscopy Sciences Supplier No. 6151507
Maximum breakage protection for known non-hazadous blood, serum, tissue, and other specimens. Accommodates  VacuTainer  Microtainer and other collection / transport tubes. Lightweight polystyrene foam construction saves postage. Reusable and recyclable. Holds 4 tubes, 110 x 19ml, 8 slides, 408 Mailing Sleeve.