Learn More
Electron Microscopy Sciences U-tek Model 443 Mini Mailers U-tek Economy Packs
SDP
Catalog No. 5033820
Encompass_Preferred
Compact protection for temperature sensitive shipments, ideal for serums, tissues, and reagents. Accommodate tubes, vials, ampoules, small bottles, and tissue flasks with space for refrigerants. Insulating, thick wall polystyrene foam construction. Holds Model 440 Mini Mailer and Model 432 U-tek refrigerant packs -23 deg C (-10 deg F).
Catalog No. 50-338-20 Supplier Electron Microscopy Sciences Supplier No. 6152143
Compact protection for temperature sensitive shipments, ideal for serums, tissues, and reagents. Accommodate tubes, vials, ampoules, small bottles, and tissue flasks with space for refrigerants. Insulating, thick wall polystyrene foam construction. Holds Model 440 Mini Mailer and Model 432 U-tek refrigerant packs -23 deg C (-10 deg F).