Learn More
Digivac Stainless Vacuum Regulator| Model 450 | Vacuum Controller for Corrosive Environments
SDP
Catalog No. NC0038949
Encompass_Preferred
The Model 450 Vacuum Controller measures 1 to 760 Torr and includes a proportional valve for vacuum regulation. Premium stainless-steel vacuum path to support use in corrosive or volatile applications. Super simple to use: just dial in your vacuum set point.Maximizes pump down speed and uses throttle vacuum control to increase pump longevity.Ideal for altitude simulation, distillations, composites research and industrial vacuum processes.Stainless steel valve and vacuum path for corrosive or hostile applications.
Catalog No. NC0038949 Supplier Digivac Supplier No. M450SS
The Model 450 Vacuum Controller measures 1 to 760 Torr and includes a proportional valve for vacuum regulation. Premium stainless-steel vacuum path to support use in corrosive or volatile applications. Super simple to use: just dial in your vacuum set point.Maximizes pump down speed and uses throttle vacuum control to increase pump longevity.Ideal for altitude simulation, distillations, composites research and industrial vacuum processes.Stainless steel valve and vacuum path for corrosive or hostile applications.