Learn More
Electron Microscopy Sciences ThermoSafe Model 480-413 Vial Shippers
SDP
Catalog No. 5033818
Encompass_Preferred
This unique array of shipping system protects frozen and refrigerated shipping vials. These ready-to- use kits include everything to keep temperature sensitive goods in top condition. Just load your product, add U-tekphase change material pack, and close the shipper. Holds 12 of 28 m.
Catalog No. 50-338-18 Supplier Electron Microscopy Sciences Supplier No. 6151913
This unique array of shipping system protects frozen and refrigerated shipping vials. These ready-to- use kits include everything to keep temperature sensitive goods in top condition. Just load your product, add U-tekphase change material pack, and close the shipper. Holds 12 of 28 m.