Learn More
Electron Microscopy Sciences ThermoSafe Insulated Shipper Model 490, EPS Box with Carton, 15 x 13.25", 16" height
SDP
Catalog No. 5033935
Encompass_Preferred
These shippers and containers offer the ideal solution for the safe storage and delivery of most refrigerated or frozen materials, including pharmaceuticals, laboratory specimens, serum and vaccines, human organs, culture packs, and nuclear and isotope materials. When shipping dry ice or utilizing dry ice as a refrigerant, these shippers provide excellent dry ice sublimation - average dry ice usage in 1 1/2 thick wall  units is 5 lbs for the first 24 hours; 3 – 4 lbs thereafter.
Type: EPS Box with Carton
Size: 15 x 13.25"
Height: 16"
Thickness: 1.5"
Catalog No. 50-339-35 Supplier Electron Microscopy Sciences Supplier No. 6153690
These shippers and containers offer the ideal solution for the safe storage and delivery of most refrigerated or frozen materials, including pharmaceuticals, laboratory specimens, serum and vaccines, human organs, culture packs, and nuclear and isotope materials. When shipping dry ice or utilizing dry ice as a refrigerant, these shippers provide excellent dry ice sublimation - average dry ice usage in 1 1/2 thick wall  units is 5 lbs for the first 24 hours; 3 – 4 lbs thereafter.
Type: EPS Box with Carton
Size: 15 x 13.25"
Height: 16"
Thickness: 1.5"