Learn More
Electron Microscopy Sciences U-tek Blue Ice Model 597, 16 oz, -23 degree C, 6½ x 6½ x ⅞ 16 OZ
SDP
Catalog No. 5033917
Encompass_Preferred
Superior protection for shipping temperature sensitive materials. Maintains critical temperatures of serums, test samples, live or frozen tissues, organs and pharmaceutical for extended periods. Laminated, metalized film exterior provides radiant heat barrier and resists punctures. U-tek -50 degree C / -58 degree F ultra low, high-tech phase change formula is ideal for products which require extreme cold but may not tolerance dry ice or CO2. Eliminate dry ice hazmat hassles.
Weight: 16 oz
Temperature: -23°C
Dimensions: 6½ x 6½ x ⅞
Catalog No. 50-339-17 Supplier Electron Microscopy Sciences Supplier No. 6153197
Superior protection for shipping temperature sensitive materials. Maintains critical temperatures of serums, test samples, live or frozen tissues, organs and pharmaceutical for extended periods. Laminated, metalized film exterior provides radiant heat barrier and resists punctures. U-tek -50 degree C / -58 degree F ultra low, high-tech phase change formula is ideal for products which require extreme cold but may not tolerance dry ice or CO2. Eliminate dry ice hazmat hassles.
Weight: 16 oz
Temperature: -23°C
Dimensions: 6½ x 6½ x ⅞