Learn More
Electron Microscopy Sciences U-tek Model 601 Mini Mailers
SDP
Catalog No. 5033825
Encompass_Preferred
Compact protection for temperature sensitive shipments, ideal for serums, tissues, and reagents. Accommodate tubes, vials, ampoules, small bottles, and tissue flasks with space for refrigerants. Insulating, thick wall polystyrene foam construction. Holds Bottles, cans, vials, cups. Can be fitted with foam insert to hold small kit components.
Catalog No. 50-338-25 Supplier Electron Microscopy Sciences Supplier No. 6152201
Compact protection for temperature sensitive shipments, ideal for serums, tissues, and reagents. Accommodate tubes, vials, ampoules, small bottles, and tissue flasks with space for refrigerants. Insulating, thick wall polystyrene foam construction. Holds Bottles, cans, vials, cups. Can be fitted with foam insert to hold small kit components.