Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences ThermoSafe Insulated Shipper Model 614, EPS Foam Box, 15.5 x 15.5", 15" height
SDP

Catalog No. 5033943
Encompass_Preferred

These shippers and containers offer the ideal solution for the safe storage and delivery of most refrigerated or frozen materials, including pharmaceuticals, laboratory specimens, serum and vaccines, human organs, culture packs, and nuclear and isotope materials. When shipping dry ice or utilizing dry ice as a refrigerant, these shippers provide excellent dry ice sublimation - average dry ice usage in 1 1/2 thick wall  units is 5 lbs for the first 24 hours; 3 – 4 lbs thereafter.
Type: EPS Foam Box
Size: 15.5 x 15.5"
Height: 15"
Thickness: 1.5"

Catalog No. 50-339-43 Supplier Electron Microscopy Sciences Supplier No. 6153614
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These shippers and containers offer the ideal solution for the safe storage and delivery of most refrigerated or frozen materials, including pharmaceuticals, laboratory specimens, serum and vaccines, human organs, culture packs, and nuclear and isotope materials. When shipping dry ice or utilizing dry ice as a refrigerant, these shippers provide excellent dry ice sublimation - average dry ice usage in 1 1/2 thick wall  units is 5 lbs for the first 24 hours; 3 – 4 lbs thereafter.
Type: EPS Foam Box
Size: 15.5 x 15.5"
Height: 15"
Thickness: 1.5"

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.