Learn More
Electron Microscopy Sciences ThermoSafe Medical Transporter Model 640
SDP
Catalog No. 5033946
Encompass_Preferred
Insulated expanded polypropylene medical transporter is ideal for on-site collection, storage, and transport of temperature sensitive specimens, medications, medical devices and organs. Durable nylon clad transporter with impact resistant expanded polyethylene container. Lightweight ergonomic design. Extra space for crushed ice and refrigerant gel pack. Washable and weather resistant. Partition divides unit, separates specimens. Built-in lid compartments.
Catalog No. 50-339-46 Supplier Electron Microscopy Sciences Supplier No. 6153740
Insulated expanded polypropylene medical transporter is ideal for on-site collection, storage, and transport of temperature sensitive specimens, medications, medical devices and organs. Durable nylon clad transporter with impact resistant expanded polyethylene container. Lightweight ergonomic design. Extra space for crushed ice and refrigerant gel pack. Washable and weather resistant. Partition divides unit, separates specimens. Built-in lid compartments.