Learn More
Electron Microscopy Sciences ThermoSafe Medical Transporter Model 641
SDP
Catalog No. 5033949
Encompass_Preferred
Insulated expanded polypropylene medical transporter is ideal for on-site collection, storage, and transport of temperature sensitive specimens, medications, medical devices and organs. Durable nylon clad transporter with impact resistant expanded polyethylene container. Lightweight ergonomic design. Extra space for crushed ice and refrigerant gel pack. Washable and weather resistant. Partitions separates specimens. Built-in lid compartments. 1/4 wall.
Catalog No. 50-339-49 Supplier Electron Microscopy Sciences Supplier No. 6153741
Insulated expanded polypropylene medical transporter is ideal for on-site collection, storage, and transport of temperature sensitive specimens, medications, medical devices and organs. Durable nylon clad transporter with impact resistant expanded polyethylene container. Lightweight ergonomic design. Extra space for crushed ice and refrigerant gel pack. Washable and weather resistant. Partitions separates specimens. Built-in lid compartments. 1/4 wall.