Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Drucker Diagnostics™ Mobile Centrifuge 642M

Catalog No. 22029587 Shop All Drucker Diagnostics Products
Change view
Click to view available options
Capacity:
6 x 10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
22-029-587 6 x 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-029-587 Supplier Drucker Diagnostics™ Supplier No. 00077009000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mobile Centrifuge that delivers high-quality, reliable blood separation every time.

  • Mobile clinical laboratory centrifuge
  • Two-button operation ensures maximum repeatability
  • Designed, built, and supported in the USA
  • Single-speed, six (6) Place centrifuge, with electronic timer preset to 10 minutes
  • For test tubes up to 10 mL or 16 x 100 mm
  • Push-button operation/safety lid-lock
  • Automatic Lid Lock – Centrifuge will not run with the lid open and the lid cannot be opened while samples are spinning
  • Positive Lid Latching – Firm mechanical latching of the lid prevents the escape of debris
  • Clear Shatter Proof Lid – Allows for safe sample observation and optical calibrations
  • Reinforced Guard Bowl – Provides maximum safety and durability
  • Brushless AC motor for smooth and quiet operation with no routine maintenance
  • Small footprint – compact design uses approximately 1 sq. ft. of counter space
  • Rotors: 3380 rpm/1600 x g
  • Includes: 6-Place Horizontal Rotor, universal tube holders, and 2 year warranty

Includes a specially designed foam car seat to ensure stability during transit. This accessory provides peace of mind and keeps your centrifuge safe and protected, even while on the go.

Specifications

Description Mobile Centrifuge for On-the-go Nurses
Includes 6-place Rotor, Tube Holders (75 to 100 mm)
Capacity 6 x 10 mL
Max. RCF 1,500 x g
Max. Speed 3,400 rpm
Refrigerated No
Noise Level 65 dB
Dimensions (L x W x H) Exterior 11.75 x 14 x 8.75 in. (29.8 x 35.6 x 22.2 cm)
Electrical Requirements 12 V - 24 V DC
Warranty 2 Years
Product Line FisherBrand
Height (English) Exterior 8.75 in.
Height (Metric) Exterior 22 cm
Length (English) Exterior 11.75 in.
Length (Metric) Exterior 30 cm
Safety Features Automatic Lid Lock, Positive Lid Latching, Clear Shatter Proof Lid, Reinforced Guard Bowl
Timer Electronic
Width (English) Exterior 14 in.
Width (Metric) Exterior 35.5 cm
Rotor 6-place Horizontal
Model 642M
Ventilated Yes
Weight (English) 11 lb.
Weight (Metric) 5 kg
For Use With (Application) Mobile Centrifugation, Home Health, Blood, Clinical Chemistry, Coag, PPP, Serum
Rotor Included Yes
Net Weight 11 lb. (5 kg)
Run Time 1 to 30 min.
Voltage 12 V
Product Type Mobile, 6-place, Fixed Angle Centrifuge
Display Type Push Button
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.