Learn More
Electron Microscopy Sciences ThermoSafe Infectious Shipper Model 660B
SDP
Catalog No. 5033929
Encompass_Preferred
These shippers are for shipping infectious agents in accordance with DOT Regulation 49CFR Parts 171 to 178 and IATA 602. They Meet US Dept of Transportation (DOT) and IATA requirement for transporting infectious substances (UN2814 and UN 2900).
Note: Compliance with UN/DOT regulations governing shipments of infectious substances in these approved shippers is contingent on the use of all supplied components. The shipper is ultimately responsible for determining the suitability of packaging for use with contents. Consult appropriate regulations. Flexible pressure vessel accepts various size tubes.
Catalog No. 50-339-29 Supplier Electron Microscopy Sciences Supplier No. 6153560B
These shippers are for shipping infectious agents in accordance with DOT Regulation 49CFR Parts 171 to 178 and IATA 602. They Meet US Dept of Transportation (DOT) and IATA requirement for transporting infectious substances (UN2814 and UN 2900).
Note: Compliance with UN/DOT regulations governing shipments of infectious substances in these approved shippers is contingent on the use of all supplied components. The shipper is ultimately responsible for determining the suitability of packaging for use with contents. Consult appropriate regulations. Flexible pressure vessel accepts various size tubes.