Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bullard™ 88VX Series Airline Respirator Accessories, Nylon Capes
SDP

Catalog No. 19021416 Shop All Bullard Company Products
Change view
Click to view available options
Overall Length (English):
28 in.
38 in.
Overall Length (Metric):
71 cm
96.5 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Overall Length (English) Overall Length (Metric)
19-021-416 28 in. 71 cm
19-021-329 38 in. 96.5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-021-416 Supplier Bullard™ Supplier No. 46VX
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Tan nylon replacement capes for use with 88VX supplied-air respirator systems.

Specifications

Color Tan
For Use With (Equipment) 88VX Series Respirator
Material Nylon
Includes Snap Hooks, Breathing Tube Strap, Additional Nylon Liner in Neck Cuff Area
Model 46VX
Overall Length (English) 28 in.
Overall Length (Metric) 71 cm
Product Type Cape
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.