Learn More
Electron Microscopy Sciences ThermoSafe Model 952 Transport Bag
SDP
Catalog No. 5033910
Encompass_Preferred
Specimen transport bags are liquid tight and tamper evident. The patented adhesive closure and continuous seal meet or exceed standards of all major air carriers and DOT. Polyethylene construction and adhesive closure provides added security during handling by personnel.
I.D.: 9 x 12"
Catalog No. 50-339-10 Supplier Electron Microscopy Sciences Supplier No. 6152552
Specimen transport bags are liquid tight and tamper evident. The patented adhesive closure and continuous seal meet or exceed standards of all major air carriers and DOT. Polyethylene construction and adhesive closure provides added security during handling by personnel.
I.D.: 9 x 12"