Learn More
Analytical Sales & Services Model C6W 6 Port Manual Injector Valve
Catalog No. 5000805016
Encompass
Model C6W 6 Port Manual Injector Valve. Six 0.016" Ports\n\nQty/Pkg: EACH
Catalog No. 50-008-05016 Supplier Analytical Sales & Services Supplier No. ADVC6W
Model C6W 6 Port Manual Injector Valve. Six 0.016" Ports\n\nQty/Pkg: EACH