Learn More
Analytical Sales & Services Model C72 Vertical 10 Port SS Injector Valve
Catalog No. 5000805778
Encompass
Model C72 Vertical 10 Port Stainless Steel Injector Valve. Rated to 15,000psi\n\nQty/Pkg: EACH
Catalog No. 50-008-05778 Supplier Analytical Sales & Services Supplier No. 8V2172
Model C72 Vertical 10 Port Stainless Steel Injector Valve. Rated to 15,000psi\n\nQty/Pkg: EACH