Learn More
Analytical Sales & Services Model C72 Vertical 6 Port SS Injector Valve
Catalog No. 5000805385
Encompass
Model C72 Vertical 6 Port Stainless Steel Injector Valve. Rated to 15,000psi
Qty/Pkg: EACH
Catalog No. 50-008-05385 Supplier Analytical Sales & Services Supplier No. 8V2672
Add to Cart
Add to Cart
Model C72 Vertical 6 Port Stainless Steel Injector Valve. Rated to 15,000psi
Qty/Pkg: EACH