Learn More
Afc International Inc Viasensor G100 CO2 Gas Detector, Lightweight Design, 0-20% Measurement Range, Unit
SDP
Catalog No. NC0543854
Encompass
The G100 CO2 Gas Detector is a portable infrared analyzer designed for precise carbon dioxide monitoring in incubators. It features advanced dual-wavelength sensor technology for quick and accurate CO2 level verification, essential in IVF and medical research. Its lightweight design, backlit LCD, and optional measurements for oxygen and humidity set it apart, ensuring optimal environmental conditions in critical applications.
Catalog No. NC0543854 Supplier Afc International Inc Supplier No. G10000L
The G100 CO2 Gas Detector is a portable infrared analyzer designed for precise carbon dioxide monitoring in incubators. It features advanced dual-wavelength sensor technology for quick and accurate CO2 level verification, essential in IVF and medical research. Its lightweight design, backlit LCD, and optional measurements for oxygen and humidity set it apart, ensuring optimal environmental conditions in critical applications.