Learn More
Electron Microscopy Sciences EMS Image Analysis Grid Pattern Model IAM-5L with Calibrated (12 pts)
SDP
Catalog No. 5034580
Encompass_Preferred
This ideal standard is for verifying or qualifying multiple image analysis parameters such as optical distortion, alignment between systems or optical paths, and cell areas for particle counting. The image area is 20 mm x 50 mm.
Catalog No. 50-345-80 Supplier Electron Microscopy Sciences Supplier No. 602165LNGC
This ideal standard is for verifying or qualifying multiple image analysis parameters such as optical distortion, alignment between systems or optical paths, and cell areas for particle counting. The image area is 20 mm x 50 mm.