Learn More
Electron Microscopy Sciences EMS Model IAM-8 Calibrated (16 pts)
SDP
Catalog No. 5034584
Encompass_Preferred
A uniqulye designed standard with 4 different grid size patterns: 0.5 x 0.5 mm grid with 25 um pitch; 1.0 x 1.0 mm grid with 50 um pitch; 2.0 x 2.0 grid with 100 um pith; and 4.0 x 4.0 mm grid with 200 um pitch. Also includes a Star Target with 15 degree angle increments. This standard can be used to test not only the overall frame distortion issue, but also linear distances for a wide variety of magnification ranges from 1x to 1000x power.
Catalog No. 50-345-84 Supplier Electron Microscopy Sciences Supplier No. 602168PGC
A uniqulye designed standard with 4 different grid size patterns: 0.5 x 0.5 mm grid with 25 um pitch; 1.0 x 1.0 mm grid with 50 um pitch; 2.0 x 2.0 grid with 100 um pith; and 4.0 x 4.0 mm grid with 200 um pitch. Also includes a Star Target with 15 degree angle increments. This standard can be used to test not only the overall frame distortion issue, but also linear distances for a wide variety of magnification ranges from 1x to 1000x power.