Learn More
Sutter Instruments Model P-2000 Laser-based puller
SDP
Catalog No. NC9973526
Encompass_Preferred
Outfitted for use with glass greater than 0.6mm outer diameter. Pullers include a glass stop, sample box of Q100-70-7.5 glass, mirrored tile and manual
Catalog No. NC9973526 Supplier Sutter Instruments Supplier No. FGP2000G
Add to Cart
Add to Cart
Request a Quote
Outfitted for use with glass greater than 0.6mm outer diameter. Pullers include a glass stop, sample box of Q100-70-7.5 glass, mirrored tile and manual