Learn More
International Crystal Model SL-3 FTIR KBr Cell, 0.5mm
Catalog No. NC1873405
Encompass_Preferred
The Model SL-3 Stainless steel sealed liquid spectrophotometer cell ensures accurate visual alignment of the cell components. These cells never have partially blocked filling ports and they never leak.
Catalog No. NC1873405 Supplier International Crystal Supplier No. 0005C872
The Model SL-3 Stainless steel sealed liquid spectrophotometer cell ensures accurate visual alignment of the cell components. These cells never have partially blocked filling ports and they never leak.