Learn More
Arrow Engineering Direct Drive Overhead Stirrer, Electric Model 1750, 120V 60HZ
Catalog No. NC3310262
Encompass_Preferred
The Arrow Mixing Products Model 1750 Electric Mixer is designed for medium-duty mixing applications, ideal for smaller batch sizes up to 20 liters. This mixer features a powerful motor with 1/14 horsepower that can achieve speeds from 50-2,000 RPM's. This model is particularly suited for applications in sectors such as; specialty chemical mixing, light industrial applications, and laboratory environments. The Model 1750 comes standard with a 3/8" dia x 12" propeller shaft, 2-1/2"dia 4 Blade propeller & aluminum coupling.
Catalog No. NC3310262 Supplier Arrow Engineering Supplier No. MODEL 1750 120VAC 60HZ/N
The Arrow Mixing Products Model 1750 Electric Mixer is designed for medium-duty mixing applications, ideal for smaller batch sizes up to 20 liters. This mixer features a powerful motor with 1/14 horsepower that can achieve speeds from 50-2,000 RPM's. This model is particularly suited for applications in sectors such as; specialty chemical mixing, light industrial applications, and laboratory environments. The Model 1750 comes standard with a 3/8" dia x 12" propeller shaft, 2-1/2"dia 4 Blade propeller & aluminum coupling.