Learn More
Arrow Engineering Direct Drive Overhead Stirrer, Electric Model 6000, 120V 60HZ
Catalog No. NC3310261
Encompass_Preferred
The Arrow Mixing Products Model 6000's compact design and light-duty capabilities make it ideal for smaller batch tasks where space and ease of use are paramount. It is designed with efficiency and reliability in mind operating at a speed range of 300-6000 RPM's and can handle batch sizes up to 20 liters of water. The Model 6000 comes standard with a 3/8" dia x 12" propeller shaft, 2-1/2"dia 4 Blade(modfied pitch) propeller & aluminum coupling.
Catalog No. NC3310261 Supplier Arrow Engineering Supplier No. MODEL 6000 120VAC 60HZ/N
The Arrow Mixing Products Model 6000's compact design and light-duty capabilities make it ideal for smaller batch tasks where space and ease of use are paramount. It is designed with efficiency and reliability in mind operating at a speed range of 300-6000 RPM's and can handle batch sizes up to 20 liters of water. The Model 6000 comes standard with a 3/8" dia x 12" propeller shaft, 2-1/2"dia 4 Blade(modfied pitch) propeller & aluminum coupling.