Learn More
Electron Microscopy Sciences Modified Hiraoka Staining Tray Space Insert, 29mm x 14mm x 4.5mm thick
SDP
Catalog No. 501931041
Encompass_Preferred
For use with the Modified Hiraoka Staining Kit. Made from polypropylene. Chemically resistant, autoclavable, and microwave safe.
Dimensions: 29mm x 14mm x 4.5mm thick
Catalog No. 50-193-1041 Supplier Electron Microscopy Sciences Supplier No. 7156031
For use with the Modified Hiraoka Staining Kit. Made from polypropylene. Chemically resistant, autoclavable, and microwave safe.
Dimensions: 29mm x 14mm x 4.5mm thick