Learn More
Labrepco Inc Modifiable Upright Freezer Rack for Standard 2″ & 3″ Boxes- Rack Only (4 Boxes Deep x 6 Boxes High)
SDP
Catalog No. NC1558073
Encompass_Preferred
These high quality Modifiable Upright Freezer Racks, including handles, are constructed of corrosion resistant stainless steel and offer convenient storage of samples in an organized format. The modifiable racks add another dimension to freezer inventory systems with the new feature of interchangeable clips to adjust the freezer racks to accommodate both 2" and 3" storage boxes.
Catalog No. NC1558073 Supplier Labrepco Inc Supplier No. UFMC4623
These high quality Modifiable Upright Freezer Racks, including handles, are constructed of corrosion resistant stainless steel and offer convenient storage of samples in an organized format. The modifiable racks add another dimension to freezer inventory systems with the new feature of interchangeable clips to adjust the freezer racks to accommodate both 2" and 3" storage boxes.